Methyl tiglate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A11964.18
A11964.30
A11964.18EA
4500
CZK
A11964.18
A11964.30
Methyl tiglate ≥98%
Methyl tiglate
Purity: 98%
CAS No.: 6622-76-0
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 39°(102°F)
Refractive Index: 1.4370
Boiling Point: 138-139°
Beilstein Reference: 1720455
Merck Reference: 14,9433
UN No.: 3272
MDL No.: MFCD00016654
Vzorec:
C₆H₁₀O₂ Bod varu: 138…139 °C Hustota: 0,950 Bod vzplanutí: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016654 CAS číslo: 6622-76-0 EINECS: 229-575-8 UN: 3272 ADR: 3,III Merck Index: 14,9433 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...