5-Bromo-2-(trifluoromethyl)pyridine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B25731.03
B25731.06
B25731.03EA
3060
CZK
B25731.03
B25731.06
5-Bromo-2-(trifluoromethyl)pyridine ≥97%
5-Bromo-2-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 436799-32-5
Molecular Formula: C6H3BrF3N
Formula Weight: 226.00
Melting Point: 39-40°
Boiling Point: 76-78°/18mm
MDL No.: MFCD06657686
Vzorec:
C₆H₃BrF₃N Bod varu: 76…78 °C (18 mmHg) Bod tání: 39…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD06657686 CAS číslo: 436799-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...