4-Methyl-5-nonanol ≥98%
Dodavatel: Thermo Scientific
B23167.09
B23167.18
B23167.30
B23167.09EA
1710
CZK
B23167.09
B23167.18
B23167.30
4-Methyl-5-nonanol ≥98%
4-Methyl-5-nonanol
Purity: 98%
CAS No.: 154170-44-2
Molecular Formula: C10H22O
Formula Weight: 158.29
Refractive Index: 1.4360
Vzorec:
C₁₀H₂₂O MW: 158,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046696 CAS číslo: 154170-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...