Tisk…
Hydroxypropyl methylcellulose (Hypromellose), 4,100 mPa.s (2% in H2O), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
M-Clarity™ quality level = MQ200
H7509-100G H7509-250G H7509-25G
SIALH7509-100GEA 9430 CZK
SIALH7509-100G SIALH7509-250G SIALH7509-25G
Hydroxypropyl methylcellulose (Hypromellose), 4,100 mPa.s (2% in H2O), Sigma-Aldrich®
Hydroxypropylmethylcellulose
Vzorec: C₃H₈O₂·xCH₄O
MDL Number: MFCD00131360
CAS číslo: 9004-65-3