Tisk…
Hydroxyethyl cellulose, M.W. 90000, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
434965-1KG 434965-250G
SIAL434965-1KGEA 4880 CZK
SIAL434965-1KG SIAL434965-250G
Hydroxyethyl cellulose, M.W. 90000, Sigma-Aldrich®
2-Hydroxyethylcellulose
MDL Number: MFCD00072770
CAS číslo: 9004-62-0