Tisk…
Hydroxyethyl cellulose, 145 mPa.s (1% in H₂0, 20 °C), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
09368-100G 09368-500G
SIAL09368-100GEA 1110 CZK
SIAL09368-100G SIAL09368-500G
Hydroxyethyl cellulose, 145 mPa.s (1% in H₂0, 20 °C), Sigma-Aldrich®
2-Hydroxyethylcellulose
MDL Number: MFCD00072770
CAS číslo: 9004-62-0