Tisk…
Hydroxyethyl cellulose, 80 - 125 mPa.s (2% in H₂0, 20 °C), Sigma-Aldrich®
Dodavatel: Merck
54290-100G 54290-500G
SIAL54290-100GEA 1310 CZK
SIAL54290-100G SIAL54290-500G
Hydroxyethyl cellulose, 80 - 125 mPa.s (2% in H₂0, 20 °C), Sigma-Aldrich®
2-Hydroxyethylcellulose
Storage Temperature: Ambient
MDL Number: MFCD00072770
CAS číslo: 9004-62-0