Tisk…

Nonaethylene glycol monododecylether (C12E9), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Nonaethylene glycol monododecyl ether, Polidocanol, 3,6,9,12,15,18,21,24,27-Nonaoxanonatriacontan-1-ol

M-Clarity™ quality level = MQ200

P9641-100G P9641-50G
SIALP9641-100GEA 8420 CZK
SIALP9641-100G SIALP9641-50G
Nonaethylene glycol monododecylether (C12E9), Sigma-Aldrich®
Nonaethylene glycol monododecylether (C12E9)
Vzorec: C₃₀H₆₂O₁₀
MW: 582.8073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043375
CAS číslo: 3055-99-0
EINECS: 221-284-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR