Tisk…

Octaethylene glycol monododecylether (C12E8), Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

P8925-1G P8925-5G
SIALP8925-1GEA 8040 CZK
SIALP8925-1G SIALP8925-5G
Octaethylene glycol monododecylether (C12E8), Sigma-Aldrich®
Octaethylene glycol monododecylether (C12E8)
Vzorec: C₂₈H₅₈O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00043234
CAS číslo: 3055-98-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR