Tisk…

Octaethylene glycol monododecylether (C12E8), Sigma-Aldrich®

Dodavatel: Merck
Varování

74680-1G 74680-25G 74680-5G
SIAL74680-1GEA 14060 CZK
SIAL74680-1G SIAL74680-25G SIAL74680-5G
Octaethylene glycol monododecylether (C12E8), Sigma-Aldrich®
Octaethylene glycol monododecylether (C12E8)
Vzorec: C₂₈H₅₈O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00043234
CAS číslo: 3055-98-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR