Tisk…

Octaethylene glycol monodecylether (C10E8), Sigma-Aldrich®

Dodavatel: Merck

74716-1ML 74716-5ML
SIAL74716-1MLEA 7040 CZK
SIAL74716-1ML SIAL74716-5ML
Octaethylene glycol monodecylether (C10E8), Sigma-Aldrich®
Octaethylene glycol monodecylether (C10E8)
Vzorec: C₂₆H₅₄O₉
CAS číslo: 24233-81-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR