Tisk…

TRITON™ X-45

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

X45-100ML X45-1L
SIALX45-100MLEA 1830 CZK
SIALX45-100ML SIALX45-1L
TRITON™ X-45
TRITON™ X-45
Storage Temperature: Ambient
MDL Number: MFCD00128254
CAS číslo: 9002-93-1
UN: 3082
ADR: 9 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR