Tisk…

L-(-)-tert-Leucine (H-Tle-OH) ≥99%

Dodavatel: Thermo Scientific

Synonyms: (S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH

L13707.03 L13707.06 L13707.14
L13707.03EA 2140 CZK
L13707.03 L13707.06 L13707.14
L-(-)-tert-Leucine (H-Tle-OH) ≥99%
L-(-)-tert-Leucine (H-Tle-OH)

Purity: 99% CAS No.: 20859-02-3 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721824

Vzorec: C₆H₁₃NO₂
MW: 131,2 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064218
CAS číslo: 20859-02-3
EINECS: 424-750-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR