Tisk…

(±)-2-Methyltetrahydrofuran-3-one ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Methyltetrahydrofuran-3-one

B20890.06 B20890.14
B20890.06EA 948 CZK
B20890.06 B20890.14
(±)-2-Methyltetrahydrofuran-3-one ≥98%
(±)-2-Methyltetrahydrofuran-3-one

Purity: 98+% CAS No.: 3188-00-9 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 39°(102°F) Refractive Index: 1.4290 Odour: Characteristic Boiling Point: 138-139° Beilstein Reference: 1341334 UN No.: 1224 MDL No.: MFCD00010423

Vzorec: C₅H₈O₂
MW: 100,12 g/mol
Bod varu: 138…139 °C
Hustota: 1,034
Bod vzplanutí: 39 °C (102 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010423
CAS číslo: 3188-00-9
EINECS: 221-685-4
UN: 1224
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR