1-Bromo-4-(ethylthio)benzene ≥97%
Dodavatel: Thermo Fisher Scientific
L13718.22
L13718.06
L13718.14
L13718.22EA
8140
CZK
L13718.22
L13718.06
L13718.14
1-Bromo-4-(ethylthio)benzene ≥97%
1-Bromo-4-(ethylthio)benzene
Purity: 97%
CAS No.: 30506-30-0
Molecular Formula: C8H9BrS
Formula Weight: 217.13
Refractive Index: 1.6069
Boiling Point: 109-111°/2mm
MDL No.: MFCD01318109
Vzorec:
C₈H₉BrS Bod varu: 109…111 °C (2 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD01318109 CAS číslo: 30506-30-0 |