Dimethyldioctadecylammonium bromide (DDA), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
D2779-10G
D2779-50G
SIALD2779-10GEA
8070
CZK
SIALD2779-10G
SIALD2779-50G
Dimethyldioctadecylammonium bromide (DDA), Sigma-Aldrich®
Dimethyldioctadecylammonium bromide (DDA)
Vzorec:
C₃₈H₈₀BrN |
MDL Number:
MFCD00041975 CAS číslo: 3700-67-2 EINECS: 223-037-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...