Tisk…

Dipentyl phthalate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Di-n-pentyl phthalate

B22910.06 B22910.14 B22910.22
B22910.06EA 2840 CZK
B22910.06 B22910.14 B22910.22
Dipentyl phthalate ≥98%
Dipentyl phthalate

Purity: 98% CAS No.: 131-18-0 Molecular Formula: C18H26O4 Formula Weight: 306.41 Flash Point: 118°(244°F) Odour: Odourless Boiling Point: 342° UN No.: 3082 MDL No.: MFCD00041934

Vzorec: C₁₈H₂₆O₄
MW: 306,41 g/mol
Bod varu: 342 °C
Hustota: 1,025
Bod vzplanutí: 118 °C (244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041934
CAS číslo: 131-18-0
EINECS: 205-017-9
UN: 3082
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR