Dipentyl phthalate 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Di-n-pentyl phthalate
357360250
357361000
357365001
ACRO357360250EA
6290
CZK
ACRO357360250
ACRO357361000
ACRO357365001
Dipentyl phthalate 97%
Dipentyl phthalate
Vzorec:
C₁₈H₂₆O₄ MW: 306,41 g/mol Bod varu: 342 °C (760 mmHg) Bod tání: –55 °C Hustota: 1,023 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00041934 CAS číslo: 131-18-0 EINECS: 205-017-9 UN: 3082 ADR: 9,III |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4885 to 1.4905 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...