7-Bromo-1-heptanol 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H54762.03
H54762.06
H54762.03EA
2110
CZK
H54762.03
H54762.06
7-Bromo-1-heptanol 96%
7-Bromo-1-heptanol
Purity: 96%
CAS No.: 10160-24-4
Molecular Formula: C7H15BrO
Formula Weight: 195.10
Flash Point: 113°(235°F)
Refractive Index: 1.4820
Boiling Point: 111-112°/4mm
MDL No.: MFCD00041708
Vzorec:
C₇H₁₅BrO MW: 195.10 g/mol Bod varu: 111…112 °C (4 mmHg) Hustota: 1.269 Bod vzplanutí: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041708 CAS číslo: 10160-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...