Tisk…

CAPS (3-(cyclohexylamino)propanesulphonic acid), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ400

C6070-100G C6070-1KG C6070-250G C6070-25G
SIALC6070-100GEA 5950 CZK
SIALC6070-100G SIALC6070-1KG SIALC6070-250G SIALC6070-25G
CAPS (3-(cyclohexylamino)propanesulphonic acid), Sigma-Aldrich®
CAPS (3-(cyclohexylamino)propanesulphonic acid)
Vzorec: C₆H₁₁NH(CH₂)₃SO₃H
Bod tání: 324 °C
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS číslo: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR