Tisk…
Aqueous/organic size exclusion chromatography columns, Asahipak GF-HQ series preparative type
Dodavatel: Resonac
F6810038 F6710020
SHOWF6810038EA 301530 CZK
SHOWF6810038 SHOWF6710020
Aqueous/organic size exclusion chromatography columns, Asahipak GF-HQ series preparative type
Kolony chromatografické
Asahipak GF-HQ series are polyvinyl alcohol-based material. Designed for both aqueous and organic solvent systems.
  • Polymer-based SEC columns exhibits solvent durability
  • Solvents used for GS-710 20F and 20G are water and methanol