Tisk…
Distribution tubes with frit for specific light solvents
Dodavatel: Lenz Laborglas GmbH & CO.KG
05432202 05432203 05432204
LENZ05432202EA 2130 CZK
LENZ05432202 LENZ05432203 LENZ05432204
Distribution tubes with frit for specific light solvents
Extraktory Extraktory Soxhlet
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Distribution tubes with frit (porosity: 0), for specific light solvents.