Tisk…
illustra Exonuclease I
Dodavatel: Cytiva
E70073Z E70073X
E70073ZEA 13640 CZK
E70073Z E70073X
illustra Exonuclease I
Enzymy
Exonuclease I acts specifically on single-stranded DNA degrading it processively in the 3'- to 5'-direction, producing 5'-mononucleotides. Applications include eliminating residual single-stranded DNA containing a 3'-terminus, measuring endonucleolytic cleavage of covalently closed circular (ccc) ssDNA, measuring DNA helicase activity.
  • 3'→5' single-stranded exonuclease
  • Inactivated by heating at 80°C for 15 minutes
  • Active in PCR buffers
  • Easily soluble in cold water and hot water
Digestion of single-stranded DNA by Exonuclease I releases deoxyribonucleoside 5'-mono-phosphates leaving 5'-terminal dinucleotides intact. DNA strands with terminal 3'-OH groups blocked by phosphoryl or acetyl groups are not removed. Monomeric 54.5 kDa bacterial exonuclease I recombinantly expressed in E. coli. One unit of the enzyme catalyzes the release of 10 nmol of acid soluble nucleotides in 30 minutes at 37°C.
Free of contaminating endonucleases, double-stranded exonucleases and ribonucleases. Greater than 95% pure as determined by SDS-PAGE.
illustra Exonuclease I, used together with illustra Alkaline Phosphatase for inactivation of dNTPs, provides effective cleanup of PCR reactions for subsequent use in DNA sequencing.
Objednat
Produktová specifikace
Název enzymu Exonuclease I
Environmentally Preferable
Zdroj
Druhy