Tisk…

Ammonia solution 20,5%

Dodavatel: VWR Collection
Nebezpečí

Synonyms: Vodný roztok amonia, Hydroxid amonný, Amoniak

VWRS83937.360EA 0 CZK
VWRS83937.360 VWRS83937.290
Ammonia solution 20,5%
Hydroxid amonný
Vzorec: NH₃
MW: 35.05 g/mol
Hustota: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS číslo: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR