Tisk…

Cocarboxylase, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Thiamine pyrophosphate chloride, Aneurinepyrophosphoric acid, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-((hydroxy(phosphonooxy)phosphoryl)oxy)ethyl)-4-methylthiazol-3-ium chloride, Cocarboxylase chloride

M-Clarity™ quality level = MQ200

C8754-5G C8754-100G C8754-1G C8754-25G
SIALC8754-5GEA 3510 CZK
SIALC8754-5G SIALC8754-100G SIALC8754-1G SIALC8754-25G
Cocarboxylase, Sigma-Aldrich®
Cocarboxylase
Vzorec: C₁₂H₁₉ClN₄O₇P₂S
MW: 460,77 g/mol
MDL Number: MFCD00038740
CAS číslo: 154-87-0
EINECS: 205-836-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR