Tisk…

Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5'-UTP 3Na, Uridine 5'-Triphosphate trisodium salt hydrate

U6750-100MG U6750-250MG U6750-500MG U6750-1G
SIALU6750-100MGEA 2480 CZK
SIALU6750-100MG SIALU6750-250MG SIALU6750-500MG SIALU6750-1G
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) hydrate, Sigma-Aldrich®
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) hydrate
Vzorec: C₉H₁₂N₂Na₃O₁₅P₃·2H₂O
Storage Temperature: Freezer
MDL Number: MFCD00044310
CAS číslo: 19817-92-6
EINECS: 243-347-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR