Tisk…

Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate, Sigma-Aldrich®

Dodavatel: Merck

94370-250MG 94370-1G
SIAL94370-250MGEA 3440 CZK
SIAL94370-250MG SIAL94370-1G
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate, Sigma-Aldrich®
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Vzorec: C₉H₁₂N₂Na₃O₁₅P₃·2H₂O
Storage Temperature: Freezer
CAS číslo: 116295-90-0
EINECS: 243-347-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR