8-Bromoguanosine hydrate ≥97%
Dodavatel: Thermo Fisher Scientific
L02992.03
L02992.06
L02992.03EA
742
CZK
L02992.03
L02992.06
8-Bromoguanosine hydrate ≥97%
8-Bromoguanosine hydrate
Purity: 97%
CAS No.: 332359-99-6
Molecular Formula: C10H12BrN5O5.xH2O
Formula Weight: 362.14(anhy)
Storage Temperature: 2°C to 8°C
Melting Point:
Vzorec:
C₁₀H₁₂BrN₅O₅·H₂O MW: 362.14 g/mol (anhy) Storage Temperature: Refrigerator |
MDL Number:
MFCD00150531 CAS číslo: 332359-99-6 EINECS: 223-677-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...