5-Bromouridine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
850187-1G
850187-250MG
SIAL850187-1GEA
3760
CZK
SIAL850187-1G
SIAL850187-250MG
5-Bromouridine, Sigma-Aldrich®
5-Bromouridine
Vzorec:
C₉H₁₁BrN₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00006528 CAS číslo: 957-75-5 EINECS: 213-486-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...