Tisk…

5-Iodo-2'-deoxyuridine (Idoxuridine), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine

I7125-25G I7125-5G
SIALI7125-25GEA 39760 CZK
SIALI7125-25G SIALI7125-5G
5-Iodo-2'-deoxyuridine (Idoxuridine), Sigma-Aldrich®
5-Iodo-2'-deoxyuridine (Idoxuridine)
Vzorec: C₉H₁₁IN₂O₅
MW: 354,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134656
CAS číslo: 54-42-2
EINECS: 200-207-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR