5-Iodo-2'-deoxyuridine (Idoxuridine) ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine
A11542.03
A11542.06
A11542.03EA
1910
CZK
A11542.03
A11542.06
5-Iodo-2'-deoxyuridine (Idoxuridine) ≥98%
5-Iodo-2'-deoxyuridine (Idoxuridine)
Purity: 98%
CAS No.: 54-42-2
Molecular Formula: C9H11IN2O5
Formula Weight: 354.10
Odour: Odourless
Melting Point: ca 190° dec.
Beilstein Reference: 30397
Merck Reference: 14,4891
MDL No.: MFCD00134656
Vzorec:
C₉H₁₁IN₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00134656 CAS číslo: 54-42-2 EINECS: 200-207-8 Merck Index: 14,4891 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...