Tisk…

Uridine, Sigma-Aldrich®

Dodavatel: Merck

U6381-25G U6381-5G
SIALU6381-25GEA 12540 CZK
SIALU6381-25G SIALU6381-5G
Uridine, Sigma-Aldrich®
Uridine
Vzorec: C₉H₁₂N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00006526
CAS číslo: 58-96-8
EINECS: 200-407-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR