Tisk…

Uridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

M-Clarity™ quality level = MQ200

U3003-50G U3003-5G
SIALU3003-50GEA 18100 CZK
SIALU3003-50G SIALU3003-5G
Uridine, Sigma-Aldrich®
Uridine
Vzorec: C₉H₁₂N₂O₆
MW: 244,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006526
CAS číslo: 58-96-8
EINECS: 200-407-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR