Tisk…

Cytidine free base, white powder

Dodavatel: MP Biomedicals

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

02101459.5 0210145910 0210145980 0210145905
ICNA02101459.5EA 1180 CZK
ICNA02101459.5 ICNA0210145910 ICNA0210145980 ICNA0210145905
Cytidine free base, white powder
Cytidine

Soluble in water.

Vzorec: C₉H₁₃N₃O₅
MW: 243,22 g/mol
Bod tání: 208…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00006545
CAS číslo: 65-46-3
EINECS: 200-610-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR