Guanosine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
M-Clarity™ quality level = MQ200
G6264-1G
G6264-25G
G6264-5G
SIALG6264-1GEA
734
CZK
SIALG6264-1G
SIALG6264-25G
SIALG6264-5G
Guanosine, Sigma-Aldrich®
Guanosine
Vzorec:
C₁₀H₁₃N₅O₅ MW: 283,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010182 CAS číslo: 118-00-3 EINECS: 204-227-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...