Tisk…

Guanosine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

M-Clarity™ quality level = MQ200

75903-100G 75903-500G
SIAL75903-100GEA 6370 CZK
SIAL75903-100G SIAL75903-500G
Guanosine, Sigma-Aldrich®
Guanosine
Vzorec: C₁₀H₁₃N₅O₅
MW: 283,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS číslo: 118-00-3
EINECS: 204-227-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR