Tisk…

Cinnamonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Phenylacrylonitrile, Cinnamonitrile

M-Clarity™ quality level = MQ100

C81004-100G C81004-25G
SIALC81004-100GEA 5170 CZK
SIALC81004-100G SIALC81004-25G
Cinnamonitrile, Sigma-Aldrich®
Cinnamonitrile
Vzorec: C₉H₇N
MW: 129,16 g/mol
MDL Number: MFCD00001930
CAS číslo: 4360-47-8
EINECS: 224-441-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR