trans-Cinnamonitrile 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(E)-3-Phenylacrylonitrile, Cinnamonitrile
B25503.22
B25503.14
B25503.22EA
4020
CZK
B25503.22
B25503.14
trans-Cinnamonitrile 97%
trans-Cinnamonitrile
Purity: 97%
CAS No.: 1885-38-7
Molecular Formula: C9H7N
Formula Weight: 129.16
Flash Point: 126°(259°F)
Refractive Index: 1.6020
Melting Point: 18-20°
Boiling Point: 253-254°
MDL No.: MFCD00001930
Vzorec:
C₉H₇N MW: 129.16 g/mol Bod varu: 253…254 °C Bod tání: 18…20 °C Hustota: 1.029 Bod vzplanutí: 126 °C (259 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001930 CAS číslo: 4360-47-8 EINECS: 217-552-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...