Tisk…

DIPSO, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid

M-Clarity™ quality level = MQ200

D0306-100G D0306-25G
SIALD0306-100GEA 26440 CZK
SIALD0306-100G SIALD0306-25G
DIPSO, Sigma-Aldrich®
DIPSO
Vzorec: C₇H₁₇NO₆S
MW: 261,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038353
CAS číslo: 68399-80-4
EINECS: 269-992-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR