Tisk…
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt
M-Clarity™ quality level = MQ200
T0807-25G T0807-5G
SIALT0807-25GEA 18080 CZK
SIALT0807-25G SIALT0807-5G
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14), Sigma-Aldrich®
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
Vzorec: C₁₉H₄₁NO₃S
MW: 363.5988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036910
CAS číslo: 14933-09-6
EINECS: 239-003-9