Tisk…
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Palmityldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, Hexadecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(palmityl)ammonio]propane-1-sulphonate, Palmityl sulphobetaine, 3-(Hexadecyldimethylammonio)propane-1-sulphonate
M-Clarity™ quality level = MQ200
H6883-25G H6883-5G
SIALH6883-25GEA 26330 CZK
SIALH6883-25G SIALH6883-5G
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16), Sigma-Aldrich®
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16)
Vzorec: C₂₁H₄₅NO₃S
MW: 391.6500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036911
CAS číslo: 2281-11-0
EINECS: 218-918-7