Betaine hydrochloride 99%, Extra pure meets analytical specification of USP
Dodavatel: Thermo Fisher Scientific
|
Varování
|
164550010
164550025
164552500
ACRO164550010EA
4820
CZK
ACRO164550010
ACRO164550025
ACRO164552500
Betaine hydrochloride 99%, Extra pure meets analytical specification of USP
Betaine hydrochloride
Vzorec:
C₅H₁₂ClNO₂ MW: 153.61 g/mol Bod tání: 241 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011903 CAS číslo: 590-46-5 EINECS: 209-683-1 |
Výsledky specifikačního testu
Appearance | Colorless to white Powder or crystals |
Infrared spectrum | Conforms (identification A) |
Water | ≤0.5 % |
Identification B | Passes test |
Residue after ignition | ≤0.1 % |
pH | 0.8 to 1.2 (1 in 4 solution) |
Titration with HClO4 | 98.5 to 100.5 % (On anhydrous basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...