Tisk…

Betaine, SAFC®

Dodavatel: Merck

Synonyms: TMG

61962-250G 61962-50G
SIAL61962-250GEA 0 CZK
SIAL61962-250G SIAL61962-50G
Betaine, SAFC®
Betaine
Vzorec: C₅H₁₁NO₂
MW: 117.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012123
CAS číslo: 107-43-7
EINECS: 203-490-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR