Betaine monohydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
B2754-1KG
B2754-500G
SIALB2754-1KGEA
11290
CZK
SIALB2754-1KG
SIALB2754-500G
Betaine monohydrate, Sigma-Aldrich®
Betaine monohydrate
Vzorec:
C₅H₁₁NO₂·1H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150010 CAS číslo: 590-47-6 EINECS: 203-490-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...