Cinnamoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Phenyl-2-propenoyl chloride, Cinnamic acid chloride
C81101-100G
C81101-500G
C81101-5G
SIALC81101-100GEA
1560
CZK
SIALC81101-100G
SIALC81101-500G
SIALC81101-5G
Cinnamoyl chloride, Sigma-Aldrich®
Cinnamoyl chloride
Vzorec:
C₉H₇ClO MW: 166,6043 g/mol |
MDL Number:
MFCD00000732 CAS číslo: 102-92-1 EINECS: 203-065-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...