Tisk…

Cinnamoyl chloride predominantly trans 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Phenyl-2-propenoyl chloride, Cinnamic acid chloride

110390050 110391000 110395000
ACRO110390050EA 1210 CZK
ACRO110390050 ACRO110391000 ACRO110395000
Cinnamoyl chloride predominantly trans 98%
Cinnamoyl chloride
Vzorec: C₉H₇ClO
Bod tání: 33…37 °C
Bod vzplanutí: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000732
CAS číslo: 102-92-1
EINECS: 203-065-5
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to yellow to orange Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 33 to 37 °C
Titration RCOCl 97.5 to 102.5 %
UV at 320 nm A: ≤0.009
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0008 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR