Polyethylene glycol dimethylether, M.W. 500 stabilizovaný, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Poly(ethylene glycol) dimethyl ether Mn 250
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
M-Clarity™ quality level = MQ200
445886-250ML
445886-50ML
SIAL445886-250MLEA
7880
CZK
SIAL445886-250ML
SIAL445886-50ML
Polyethylene glycol dimethylether, M.W. 500 stabilizovaný, Sigma-Aldrich®
Polyethylenglycoldimethylether
Vzorec:
H₃C(OCH₂CH₂)nOCH₃ |
MDL Number:
MFCD00147972 CAS číslo: 24991-55-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...