Tisk…
Polyethylene glycol dimethylether, M.W. 2,000, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Poly(ethylene glycol) dimethyl ether Mn 250
M-Clarity™ quality level = MQ200
445908-250G 445908-50G
SIAL445908-250GEA 7420 CZK
SIAL445908-250G SIAL445908-50G
Polyethylene glycol dimethylether, M.W. 2,000, Sigma-Aldrich®
Polyethylenglycoldimethylether
Vzorec: H₃C(OCH₂CH₂)nOCH₃
MDL Number: MFCD00147972
CAS číslo: 24991-55-7