Polyethylene glycol dimethylether, M.W. 1,000, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Poly(ethylene glycol) dimethyl ether Mn 250
M-Clarity™ quality level = MQ100
445894-250G
445894-50G
SIAL445894-250GEA
7810
CZK
SIAL445894-250G
SIAL445894-50G
Polyethylene glycol dimethylether, M.W. 1,000, Sigma-Aldrich®
Polyethylenglycoldimethylether
Vzorec:
H₃C(OCH₂CH₂)nOCH₃ |
MDL Number:
MFCD00147972 CAS číslo: 24991-55-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...