Tisk…

1-Vinyl-2-pyrrolidone stabilizovaný for synthesis, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: N-Vinyl-2-pyrrolidone, 1-Vinyl-2-pyrrolidinone, N-Vinyl-2-pyrrolidinone

Stabilization: Stabilised with N,N'-di-sec-butyl-p-phenylenediamine
M-Clarity™ quality level = MQ200

8.08518.0005 8.08518.0250
8.08518.0005EA 887 CZK
8.08518.0005 8.08518.0250
1-Vinyl-2-pyrrolidone stabilizovaný for synthesis, Sigma-Aldrich®
1-Vinyl-2-pyrrolidone
Vzorec: C₆H₉NO
MW: 111,1418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003197
CAS číslo: 88-12-0
EINECS: 201-800-4
UN: 2810
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Solubility (20 °C) soluble
Melting Point 13 - 14 °C
Molar Mass 111.14 g/mol
Boiling Point 147 - 149 °C (113 hPa)
Vapor Pressure 0.12 hPa (20 °C)
Flash Point 95 °C
Refractive Index 1.512 (20 °C, 589 nm)
Explosion Limit 1.4 - 10 %(V)
Density 1.04 g/cm3 (20 °C)
pH 9 - 10 (100 g/l, H2O, 20 °C)
Ignition Point 240 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR